Merck
  • Comparison of counter-current chromatography and preparative high performance liquid chromatography applied to separating minor impurities in drug preparations.
Comparison of counter-current chromatography and preparative high performance liquid chromatography applied to separating minor impurities in drug preparations.
Journal of chromatography. A (2014-04-29)
Shucai Li, Wenwen Wang, Huan Tang, Kai Chen, Jianhong Yang, Linhong He, Haoyu Ye, Aihua Peng, Lijuan Chen
ABSTRAKT
Drug impurity profiling and identification are carried out along with the drug discovery process. Due to its inherent low concentration in drug products, the isolation and purification of impurities present a challenge to drug development processes. In our development of honokiol and quercetin as anticancer drug candidates, counter-current chromatography (CCC) and preparative HPLC were used for the impurity profiling and identification of honokiol and quercetin. Several performance parameters such as separation column volume, maximum sample loading, separation time, solvent consumption and sample throughput were investigated in order to compare the separation efficiency. We found that the sample loading capacity and therefore the throughput of preparative HPLC were not satisfactory, while CCC provided larger sample loading (especially for a sample with poor solubility), consumed less solvent and produced higher throughput than preparative HPLC. Six impurities of honokiol including one new compound were isolated in the present work.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Honokiol, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Honokiol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%