Merck
Leucine/isoleucine zipper coordination of ion channel macromolecular signaling complexes in the heart. Roles in inherited arrhythmias.
Trends in cardiovascular medicine (2003-02-15)
R S Kass, J Kurokawa, S O Marx, A R Marks
ABSTRAKT
The sympathetic nervous system controls the force and rate of contraction of the heart. The rapid response to stress and exercise mediated by increased sympathetic nervous system (SNS) activity requires the coordinated regulation of several ion channels in response to activation of beta-adrenergic receptors. The microenvironment of target channels is mediated by the assembly of macromolecular signaling complexes in which targeting proteins recruit phosphatases and kinases and in turn bind directly to the channel protein via highly conserved leucine/isoleucine zippers (LIZs). Disruption of local signaling by disease-associated LIZ mutations unbalances the physiologic response to SNS stimulation and increases the risk of arrhythmia in mutation carriers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Isoleucine, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Isoleucine
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)