Merck
In defense of magnesium sulfate.
Obstetrics and gynecology (2009-05-23)
John P Elliott, David F Lewis, John C Morrison, Thomas J Garite
ABSTRAKT
Magnesium sulfate has been used by obstetricians for more than 25 years to treat preterm labor. Magnesium sulfate is effective in delaying delivery for at least 48 hours in patients with preterm labor when used in higher dosages. There do not seem to be any harmful effects of the drug on the fetus, and indeed there is a neuroprotective effect in reducing the incidence of cerebral palsy in premature newborns weighing less than 1,500 g.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis