Merck
Acanthosis nigricans in the setting of niacin therapy.
Dermatology online journal (2011-10-28)
Rachael Hartman, Taylor Defelice, Julia Tzu, Shane Meehan, Miguel Sanchez
ABSTRAKT
We report the case of a 63-year-old obese man with a rapid-onset of widespread acanthosis nigricans (AN) in the setting of having recently initiated treatment with niacin for dyslipidemia. Although obesity and insulin-resistance are risk factors for AN, AN associated with endocrine dysfunction tends to have a more gradual onset and limited involvement. Owing to our patient's age, the rapid onset, and extensive distribution of his eruption, we initially were concerned about paraneoplastic AN. However, an evaluation for a malignant condition was negative. The timing of the onset of our patient's eruption within several months of starting niacin therapy is consistent with niacin-induced AN. Niacin is known to cause rapidly progressive, widespread AN that is reversible upon discontinuation of the medication. We discuss the pathogenesis of AN, which is thought to be the final common manifestation of stimulation of different subtypes of tyrosine kinase receptors by various epidermal growth factors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacyna, United States Pharmacopeia (USP) Reference Standard
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard