Merck
Vitamin B12 and ageing: current issues and interaction with folate.
Annals of clinical biochemistry (2013-04-18)
Catherine F Hughes, Mary Ward, Leane Hoey, Helene McNulty
ABSTRAKT
A compromised vitamin B12 status is common in older people despite dietary intakes that typically far exceed current recommendations. The maintenance of an optimal status of vitamin B12 is not only dependent on adequate dietary intake but more critically on effective absorption which diminishes with age. The measurement of vitamin B12 is complicated by the lack of a gold standard assay. There are a number of direct and functional indicators of vitamin B12 status; however, none of these are without limitations and should be used in combination. Vitamin B12 is of public health importance, not only because deficiency leads to megaloblastic anaemia and irreversible nerve damage, but also because emerging evidence links low B12 to an increased risk of a number of age-related diseases, including cardiovascular disease, cognitive dysfunction, dementia and osteoporosis. Furthermore, there are concerns relating to potential adverse effects for older adults with low vitamin B12 status of over-exposure to folic acid in countries where there is mandatory fortification of food with folic acid. The aim of this review is to examine the known and emerging issues related to vitamin B12 in ageing, its assessment and inter-relationship with folate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®