Merck
Vinyl chloride mechanistic data and risk assessment: DNA reactivity and cross-species quantitative risk extrapolation.
Pharmacology & therapeutics (1996-01-01)
J Whysner, C C Conaway, L Verna, G M Williams
ABSTRAKT
Vinyl chloride produced several tumor types among species. Angiosarcoma of the liver is found in all tested species, including humans with occupational exposures. Vinyl chloride is biotransformed by CYP2E1 to DNA-reactive chloroethylene oxide producing cyclic etheno adducts, which are mutagenic. The dose-response for angiosarcoma of the liver formation in rodents is supralinear, which is consistent with saturation of metabolic activation, and the tumor rate in humans at occupational exposure levels is similar to that for equivalent exposures in rodents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol
Informacje o cenach i dostępności nie są obecnie dostępne.