Merck
Psychological stress and corticotropin-releasing hormone increase intestinal permeability in humans by a mast cell-dependent mechanism.
Gut (2013-10-25)
Tim Vanuytsel, Sander van Wanrooy, Hanne Vanheel, Christophe Vanormelingen, Sofie Verschueren, Els Houben, Shadea Salim Rasoel, Joran Tόth, Lieselot Holvoet, Ricard Farré, Lukas Van Oudenhove, Guy Boeckxstaens, Kristin Verbeke, Jan Tack
ABSTRAKT
Intestinal permeability and psychological stress have been implicated in the pathophysiology of IBD and IBS. Studies in animals suggest that stress increases permeability via corticotropin-releasing hormone (CRH)-mediated mast cell activation. Our aim was to investigate the effect of stress on intestinal permeability in humans and its underlying mechanisms. Small intestinal permeability was quantified by a 2 h lactulose-mannitol urinary excretion test. In a first study, 23 healthy volunteers were subjected to four different conditions: control; indomethacin; public speech and anticipation of electroshocks. In a second study, five test conditions were investigated in 13 volunteers: control; after pretreatment with disodium cromoglycate (DSCG); administration of CRH; DSCG+CRH and DSCG+public speech. Indomethacin, as a positive comparator (0.071±0.040 vs 0.030±0.022; p<0.0001), and public speech (0.059±0.040; p<0.01), but not the shock protocol increased intestinal permeability. Similarly, salivary cortisol was only increased after public speech. Subgroup analysis demonstrated that the effect of public speech on permeability was only present in subjects with a significant elevation of cortisol. CRH increased the lactulose-mannitol ratio (0.042±0.021 vs 0.028±0.009; p=0.02), which was inhibited by the mast cell stabiliser DSCG. Finally, intestinal permeability was unaltered by public speech with DSCG pretreatment. Acute psychological stress increases small intestinal permeability in humans. Peripheral CRH reproduces the effect of stress and DSCG blocks the effect of both stress and CRH, suggesting the involvement of mast cells. These findings provide new insight into the complex interplay between the central nervous system and GI function in man.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Cromolyn sodium salt, ≥95%
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Lactulose for peak identification, European Pharmacopoeia (EP) Reference Standard
Sodium cromoglicate, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard