Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Silver, insulated wire, 5m, conductor diameter 0.075mm, insulation thickness 0.010mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, microfoil, disks, 25mm, thinness 0.1μm, specific density 110μg/cm2, permanent mylar 3.5μm support, 99.95+%
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%
Silver, insulated wire, 50m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Silver, rod, 50mm, diameter 8.0mm, as drawn, 99.95+%
Silver, insulated wire, 5m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Silver, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.015mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, tube, 1000mm, outside diameter 4.65mm, inside diameter 4.15mm, wall thickness 0.25mm, as drawn, 99.95+%
Silver, microfoil, disks, 10mm, thinness 0.5μm, specific density 522.6μg/cm2, permanent mylar 3.5μm support, 99.95+%
Silver, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%
Silver, insulated wire, 2m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Silver, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.015mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, rod, 1000mm, diameter 3.5mm, as drawn, 99.95+%
Silver, tube, 100mm, outside diameter 4.65mm, inside diameter 4.15mm, wall thickness 0.25mm, as drawn, 99.95+%
Silver, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%
Silver, tube, 200mm, outside diameter 1.75mm, inside diameter 1.55mm, wall thickness 0.1mm, as drawn, 99.95+%