Merck
Survival improvement of poor-prognosis AML/MDS patients by maintenance treatment with low-dose chemotherapy and differentiating agents.
Annals of hematology (2014-04-08)
Dario Ferrero, Elena Crisà, Filippo Marmont, Ernesta Audisio, Chiara Frairia, Valentina Giai, Tiziana Gatti, Moreno Festuccia, Benedetto Bruno, Ludovica Riera, Roberto Passera, Mario Boccadoro
ABSTRAKT
We evaluated a maintenance, post-remission treatment with low-dose chemotherapy plus differentiating agents on poor-prognosis acute myeloid leukemia (AML)/myelodysplastic syndrome (MDS) patients ineligible to allografting. Patients had either age over 60 and/or secondary AML, therapy-related AML, previous relapse, high-risk MDS. Forty-five patients received the maintenance therapy based on two alternated schedules: (a) 6-thioguanine + 13-cis retinoic acid + dihydroxylated vitamin D3 and (b) low-dose cytarabine + 6-mercaptopurine + all-trans retinoic acid + dihydroxylated vitamin D3. We compared their outcome, at a median follow-up of 52 months, to that of a matched population of 49 patients who stopped treatments after consolidation. Maintenance group had a lower relapse incidence (70.3 vs. 86.4 % at 5 years p = 0.007) and a longer disease-free survival (median 21.2 vs. 8.7 months, p = 0.017). The relapse reduction improved overall survival: median 40.4 months (35.9 % at 5 years) for maintenance group vs. 15.8 (14.2 % at 5 years) for controls (p = 0.005). At multivariate Cox analysis, both cytogenetic and maintenance therapies resulted independent outcome predictors for overall survival. Maintenance treatment also reduced minimal residual disease (detected by WT1 and CBFβ-MYH11) in five of eight evaluable patients. The present results suggest that our strategy of maintenance therapy might improve the outcome of poor-risk AML/MDS patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Mitoxantrone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Adenine 9-β-D-arabinofuranoside, ≥99%
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder