Merck
[Vaginal bleeding as symptom of a medical emergency--pitfalls of therapy with new oral anticoagulants - case 5/2014].
Deutsche medizinische Wochenschrift (1946) (2014-07-30)
Dorothea Baumann, Michael Haap, Christos Tsaousidis, Bernhard Krämer, Jürgen Schreieck, Karl Jaschonek, Ingo Rettig, Andreas Peter, Diethelm Wallwiener, Ferruh Artunc
ABSTRAKT
A 90-year-old female patient treated with dabigatran for atrial fibrillation presented emergently with a spontaneous vaginal bleeding due to endometrium carcinoma. Laboratory analysis revealed azotemia consistent with acute-on-chronic renal failure. Coagulation was deranged (prolongation of activated partial thromboplastin time (aPTT) by 3.5-fold, international normalized ratio (INR) 7.8) due to a massive accumulation of dabigatran (measured plasma concentration 2230 ng/ml). Vaginal bleedings were treated with external tamponade. Hemodialysis treatment was commenced due to uremia and dabigatran accumulation. Over night, the patient was dialysed with a SLED (sustained low efficiency dialysis) regimen. After volume resuscitation renal function promptly ensued indicating prerenal azotemia as a cause of renal failure. After two more hemodialysis sessions dabigatran concentrations were no longer detecable and this was paralleled by normalization of coagulation tests. Dabigatran can accumulate massively during acute renal failure and evoke a life-threatening bleeding diathesis. Dabigatran can be removed with low efficient dialysis (such as SLED).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Alanine, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
β-Alanine, 99%
Supelco
β-Alanine, analytical standard
Sigma-Aldrich
β-Alanine, BioUltra, ≥99.0% (NT)
Supelco
Beta Alanine, Pharmaceutical Secondary Standard; Certified Reference Material