Merck
Detection of hepatotoxicity potential with metabolite profiling (metabolomics) of rat plasma.
Toxicology letters (2014-08-03)
W Mattes, K Davis, E Fabian, J Greenhaw, M Herold, R Looser, W Mellert, S Groeters, H Marxfeld, N Moeller, G Montoya-Parra, A Prokoudine, B van Ravenzwaay, V Strauss, T Walk, H Kamp
ABSTRAKT
While conventional parameters used to detect hepatotoxicity in drug safety assessment studies are generally informative, the need remains for parameters that can detect the potential for hepatotoxicity at lower doses and/or at earlier time points. Previous work has shown that metabolite profiling (metabonomics/metabolomics) can detect signals of potential hepatotoxicity in rats treated with doxorubicin at doses that do not elicit hepatotoxicity as monitored with conventional parameters. The current study extended this observation to the question of whether such signals could be detected in rats treated with compounds that can elicit hepatotoxicity in humans (i.e., drug-induced liver injury, DILI) but have not been reported to do so in rats. Nine compounds were selected on the basis of their known DILI potential, with six other compounds chosen as negative for DILI potential. A database of rat plasma metabolite profiles, MetaMap(®)Tox (developed by metanomics GmbH and BASF SE) was used for both metabolite profiles and mode of action (MoA) metabolite signatures for a number of known toxicities. Eight of the nine compounds with DILI potential elicited metabolite profiles that matched with MoA patterns of various rat liver toxicities, including cholestasis, oxidative stress, acetaminophen-type toxicity and peroxisome proliferation. By contrast, only one of the six non-DILI compounds showed a weak match with rat liver toxicity. These results suggest that metabolite profiling may indeed have promise to detect signals of hepatotoxicity in rats treated with compounds having DILI potential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Propyl-2-thiouracil, enzyme inhibitor
USP
Captopril, United States Pharmacopeia (USP) Reference Standard
USP
Fenytoina, United States Pharmacopeia (USP) Reference Standard
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenytoin, Pharmaceutical Secondary Standard; Certified Reference Material
Phenytoin, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Captopril, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Atropine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Captopril, meets USP testing specifications
Supelco
Phenytoin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
6-Propyl-2-thiouracil, VETRANAL®, analytical standard
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Phenytoin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Zidovudine, European Pharmacopoeia (EP) Reference Standard
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Zidovudine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Methotrexate
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Atropine, European Pharmacopoeia (EP) Reference Standard