Merck
Tumoral RANKL activates astrocytes that promote glioma cell invasion through cytokine signaling.
Cancer letters (2014-08-01)
Jun-Kyum Kim, Xiong Jin, Young-Woo Sohn, Xun Jin, Hee-Young Jeon, Eun-Jung Kim, Seok Won Ham, Hye-Min Jeon, So-Young Chang, Se-Yeong Oh, Jinlong Yin, Sung-Hak Kim, Jong Bae Park, Ichiro Nakano, Hyunggee Kim
ABSTRAKT
The invasiveness of glioblastoma is a major cause of poor prognosis and relapse. However, the molecular mechanism controlling glioma cell invasion is poorly understood. Here, we report that receptor activator of nuclear factor kappa-B (NFκB) ligand (RANKL) promotes glioma cell invasion in vivo, but not in vitro. Unlike the invasiveness under in vitro culture conditions, in vivo xenograft studies revealed that LN229 cells expressing high endogenous RANKL generated more invasive tumors than U87MG cells expressing relatively low endogenous RANKL. Consistently, RANKL-overexpressing U87MG resulted in invasive tumors, whereas RANKL-depleted LN229 generated rarely invasive tumors. We found that the number of activated astrocytes was markedly increased in the periphery of RANKL-high invasive tumors. RANKL activated astrocytes through NFκB signaling and these astrocytes in turn secreted various factors which regulate glioma cell invasion. Among them, transforming growth factor β (TGF-β) signaling was markedly increased in glioblastoma specimens and xenograft tumors expressing high levels of RANKL. These results indicate that RANKL contributes to glioma invasion by modulating the peripheral microenvironment of the tumor, and that targeting RANKL signaling has important implications for the prevention of highly invasive glioblastoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
RANK Ligand/TRANCE human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
sRANKL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%