Merck
Enhancement of Runx2 expression is potentially linked to β-catenin accumulation in canine intervertebral disc degeneration.
Journal of cellular physiology (2014-06-12)
Munetaka Iwata, Takeshi Aikawa, Takaharu Hakozaki, Kiyotaka Arai, Hiroki Ochi, Hirotaka Haro, Masahiro Tagawa, Yoshinori Asou, Yasushi Hara
ABSTRAKT
Intervertebral disc degeneration (IVDD) greatly affects the quality of life. The nucleus pulposus (NP) of chondrodystrophic dog breeds (CDBs) is similar to the human NP because the cells disappear with age and are replaced by fibrochondrocyte-like cells. Because IVDD develops as early as within the first year of life, we used canines as a model to investigate the in vitro mechanisms underlying IVDD. The mechanism underlying age-related IVDD, however, is poorly understood. Several research groups have suggested that Wnt/β-catenin signaling plays an important role in IVDD. However, the role of Wnt/β-catenin signals in IVD cells is not yet well understood. Here, we demonstrate that Wnt/β-catenin signaling could enhance Runx2 expression in IVDD and lead to IVD calcification. Nucleus pulposus (NP) tissue was obtained from Beagle dogs after evaluation of the degeneration based on magnetic resonance imaging (MRI). Histological analysis showed that lack of Safranin-O staining, calcified area, and matrix metalloproteinase (MMP) 13-positive cells increased with progression of the degeneration. Furthermore, the levels of β-catenin- and Runx2-positive cells also increased. Real-time reverse-transcription polymerase chain reaction analysis showed that the MRI signal intensity and mRNA expression levels of β-catenin and Runx2 are correlated in NP tissues. Moreover, supplementation of LiCl induced β-catenin accumulation and Runx2 expression. In contrast, FH535 inhibited LiCl-induced upregulation. These results suggest that Runx2 transcript and protein expression, potentially in combination with β-catenin accumulation, are enhanced in degenerated and calcified intervertebral discs of CDBs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Lithium chloride solution, 2 M in ethanol
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Supelco
Roztwór elektrolitu, nonaqueous, LiCl in ethanol (saturated)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol