Merck
Environmental applications of chitosan and its derivatives.
Reviews of environmental contamination and toxicology (2014-11-05)
Soon Kong Yong, Manoj Shrivastava, Prashant Srivastava, Anitha Kunhikrishnan, Nanthi Bolan
ABSTRAKT
Chitosan originates from the seafood processing industry and is one of the most abundant of bio-waste materials. Chitosan is a by-product of the alkaline deacetylation process of chitin. Chemically, chitosan is a polysaccharide that is soluble in acidic solution and precipitates at higher pHs. It has great potential for certain environmental applications, such as remediation of organic and inorganic contaminants, including toxic metals and dyes in soil, sediment and water, and development of contaminant sensors. Traditionally, seafood waste has been the primary source of chitin. More recently, alternative sources have emerged such as fungal mycelium, mushroom and krill wastes, and these new sources of chitin and chitosan may overcome seasonal supply limitations that have existed. The production of chitosan from the above-mentioned waste streams not only reduces waste volume, but alleviates pressure on landfills to which the waste would otherwise go. Chitosan production involves four major steps, viz., deproteination, demineralization, bleaching and deacetylation. These four processes require excessive usage of strong alkali at different stages, and drives chitosan's production cost up, potentially making the application of high-grade chitosan for commercial remediation untenable. Alternate chitosan processing techniques, such as microbial or enzymatic processes, may become more cost-effective due to lower energy consumption and waste generation. Chitosan has proved to be versatile for so many environmental applications, because it possesses certain key functional groups, including - OH and -NH2 . However, the efficacy of chitosan is diminished at low pH because of its increased solubility and instability. These deficiencies can be overcome by modifying chitosan's structure via crosslinking. Such modification not only enhances the structural stability of chitosan under low pH conditions, but also improves its physicochemical characteristics, such as porosity, hydraulic conductivity, permeability, surface area and sorption capacity. Crosslinked chitosan is an excellent sorbent for trace metals especially because of the high flexibility of its structural stability. Sorption of trace metals by chitosan is selective and independent of the size and hardness of metal ions, or the physical form of chitosan (e.g., film, powder and solution). Both -OH and -NH2 groups in chitosan provide vital binding sites for complexing metal cations. At low pH, -NH3 + groups attract and coagulate negatively charged contaminants such as metal oxyanions, humic acids and dye molecules. Grafting certain functional molecules into the chitin structure improves sorption capacity and selectivity for remediating specific metal ions. For example, introducing sulfur and nitrogen donor ligands to chitosan alters the sorption preference for metals. Low molecular weight chitosan derivatives have been used to remediate metal contaminated soil and sediments. They have also been applied in permeable reactive barriers to remediate metals in soil and groundwater. Both chitosan and modified chitosan have been used to phytoremediate metals; however, the mechanisms by which they assist in mobilizing metals are not yet well understood. In addition, microbes have been used in combination with chitosan to remediate metals (e.g., Cu and Zn) in contaminated soils. Chitosan has also been used to remediate organic contaminants, such as oil-based wastewater, dyes, tannins, humic acids, phenols, bisphenoi-A, p-benzoquinone, organo-phosphorus insecticides, among others. Chitosan has also been utilized to develop optical and electrochemical sensors for in-situ detection of trace contaminants. In sensor technology, naturally-derived chitosan is used primarily as an immobilizing agent that results from its enzyme compatibility, and stabilizing effect on nanoparticles. Contaminant-sensing agents, such as enzymes, microbes and nanoparticles, have been homogeneously immobilized in chitosan gels by using coagulating (e.g., alginate, phosphate) or crosslinking agents (e.g., GA, ECH). Such immobilization maintains the stability of sensing elements in the chitosan gel phase, and prevents inactivation and loss of the sensing agent. In this review, we have shown that chitosan, an efficient by-product of a waste biomaterial, has great potential for many environmental applications. With certain limitations, chitosan and its derivatives can be used for remediating contaminated soil and wastewater. Notwithstanding, further research is needed to enhance the physicochemical properties of chitosan and mitigate its deficiencies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Chitosan, low molecular weight
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Chitosan, high molecular weight
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Chitosan, medium molecular weight
Informacje o cenach i dostępności nie są obecnie dostępne.