Merck
Low-grade glioma.
Cancer treatment and research (2014-12-04)
Priya Kumthekar, Jeffrey Raizer, Simran Singh
ABSTRAKT
Low-grade gliomas are slower growing than their high-grade counterparts. They account for 10-20 % of all primary brain tumors. Median survival is between 4.7 and 9.8 years. The goal of treatment is to prolong overall survival while maintaining good quality of life (QOL). Recent data favors early surgical resection. EOR is associated with delayed tumor recurrence and improved survival. Additional therapy with chemotherapy or radiation is indicated in patients with high-risk features. Lower doses (between 45 and 50.4 Gy) have been shown to be as effective without adverse effects compared to higher doses. Recent trials have shown benefit in combining chemotherapy with radiation compared to radiation alone. The optimal chemotherapeutic regimen (PCV or temozolomide (TMZ)) remains unknown, although TMZ is easier to administer and better tolerated by patients. Novel molecular markers including 1p/19q chromosomal codeletion and isocitrate dehydrogenase 1 (IDH1) mutation have been correlated with treatment response and survival.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in Sf9 cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in E. coli, lyophilized powder, ≥80 units/mg protein
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type I, 0.5-3.0 unit/mg solid (plus numerous enzyme activities associated with porcine heart)