Merck
The shikimate dehydrogenase family: functional diversity within a conserved structural and mechanistic framework.
Archives of biochemistry and biophysics (2014-12-20)
James Peek, Dinesh Christendat
ABSTRAKT
Shikimate dehydrogenase (SDH) catalyzes the NADPH-dependent reduction of 3-deydroshikimate to shikimate, an essential reaction in the biosynthesis of the aromatic amino acids and a large number of other secondary metabolites in plants and microbes. The indispensible nature of this enzyme makes it a potential target for herbicides and antimicrobials. SDH is the archetypal member of a large protein family, which contains at least four additional functional classes with diverse metabolic roles. The different members of the SDH family share a highly similar three-dimensional structure and utilize a conserved catalytic mechanism, but exhibit distinct substrate preferences, making the family a particularly interesting system for studying modes of substrate recognition used by enzymes. Here, we review our current understanding of the biochemical and structural properties of each of the five previously identified SDH family functional classes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Shikimic acid, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Supelco
Shikimic acid, analytical standard