Merck
Assessment of emergency responders after a vinyl chloride release from a train derailment - New Jersey, 2012.
MMWR. Morbidity and mortality weekly report (2015-01-13)
Kimberly Brinker, Margaret Lumia, Karl V Markiewicz, Mary Anne Duncan, Chad Dowell, Araceli Rey, Jason Wilken, Alice Shumate, Jamille Taylor, Renée Funk
ABSTRAKT
On November 30, 2012, at approximately 7:00 am, a freight train derailed near a small town in New Jersey. Four tank cars, including a breached tank car carrying vinyl chloride, landed in a tidal creek. Vinyl chloride, a colorless gas with a mild, sweet odor, is used in plastics manufacture. Acute exposure can cause respiratory irritation and headache, drowsiness, and dizziness; chronic occupational exposure can result in liver damage, accumulation of fat in the liver, and tumors (including angiosarcoma of the liver). Because health effects associated with acute exposures have not been well studied, the New Jersey Department of Health requested assistance from the Agency for Toxic Substances and Disease Registry (ATSDR) and CDC. On December 11, teams from these agencies deployed to assist the New Jersey Department of Health in conducting an assessment of exposures in the community as well as the occupational health and safety of emergency personnel who responded to the incident. This report describes the results of the investigation of emergency personnel. A survey of 93 emergency responders found that 26% of respondents experienced headache and upper respiratory symptoms during the response. A minority (22%) reported using respiratory protection during the incident. Twenty-one (23%) of 92 respondents sought medical evaluation. Based on these findings, CDC recommended that response agencies 1) implement the Emergency Responder Health Monitoring and Surveillance (ERHMS) system for ongoing health monitoring of the emergency responders involved in the train derailment response and 2) ensure that in future incidents, respiratory protection is used when exposure levels are unknown or above the established occupational exposure limits.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol
Informacje o cenach i dostępności nie są obecnie dostępne.