Merck
Significant association of MTHFD1 1958G>A single nucleotide polymorphism with nonsyndromic cleft lip and palate in Indian population.
Medicina oral, patologia oral y cirugia bucal (2014-08-19)
Jyotsna Murthy, Venkatesh-Babu Gurramkonda, Bhaskar V K S Lakkakula
ABSTRAKT
Nonsyndromic cleft lip and palate (NSCLP) is genetically distinct from those with syndromic clefts, and accounts for ~70% of cases with Oral clefts. Folate, or vitamin B9, is an essential nutrient in our diet. Allelic variants in genes involved in the folate pathway might be expected to have an impact on risk of oral clefts. Given the key role of methylenetetrahydrofolate dehydrogenase 1 (MTHFD1) in folate metabolism, it would be of significant interest to assess its role in NSCLP etiology. The present study aims at examining the association between MTHFD1 1958G>A polymorphism and NSCLP risk by conducting a case-control study in south Indian population. Our sample comprised of 142 cases with nonsyndromic clefts and 141 controls without clefts or family history of clefting. The MTHFD1 1958G>A polymorphism was genotyped using PCR-RFLP. An increased risk was found for the heterozygous 1958GA (OR=2.44; P=0.020) and homozygous 1958AA (OR=2.45; P=0.012) genotypes in the children. When the dominant model (AG+AA vs GG) was applied the risk remained the same as co-dominant model, but the level of significance increased (OR=2.44; P=0.002). The results indicated the MTHFD1 1958G>A polymorphism to be one of the important genetic determinants of NSCLP risk in South Indian subjects.