Merck
  • Opposing effects of low molecular weight heparins on the release of inflammatory cytokines from peripheral blood mononuclear cells of asthmatics.
Opposing effects of low molecular weight heparins on the release of inflammatory cytokines from peripheral blood mononuclear cells of asthmatics.
PloS one (2015-03-05)
Madhur D Shastri, Niall Stewart, Mathew Eapen, Gregory M Peterson, Syed Tabish R Zaidi, Nuri Gueven, Sukhwinder Singh Sohal, Rahul P Patel
ABSTRAKT
T-cell-mediated inflammatory cytokines, such as interleukin (IL)-4, IL-5, IL-13 and tumor necrosis factor-alpha (TNF-α), play an important role in the initiation and progression of inflammatory airways diseases. Low-molecular-weight heparins (LMWHs), widely used anticoagulants, possess anti-inflammatory properties making them potential treatment options for inflammatory diseases, including asthma. In the current study, we investigated the modulating effects of two LMWHs (enoxaparin and dalteparin) on the release of cytokines from stimulated peripheral blood mononuclear cells (PBMCs) of asthmatic subjects to identify the specific components responsible for the effects. PBMCs from asthmatic subjects (consist of ~75% of T-cells) were isolated from blood taken from ten asthmatic subjects. The PBMCs were pre-treated in the presence or absence of different concentrations of LMWHs, and were then stimulated by phytohaemagglutinin for the release of IL-4, IL-5, IL-13 and TNF-α. LMWHs were completely or selectively desulfated and their anticoagulant effect, as well as the ability to modulate cytokine release, was determined. LMWHs were chromatographically fractionated and each fraction was tested for molecular weight determination along with an assessment of anticoagulant potency and effect on cytokine release. Enoxaparin inhibited cytokine release by more than 48%, whereas dalteparin increased their release by more than 25%. The observed anti-inflammatory effects of enoxaparin were independent of their anticoagulant activities. Smaller fractions, in particular dp4 (four saccharide units), were responsible for the inhibitory effect of enoxaparin. Whereas, the larger fractions, in particular dp22 (twenty two saccharide units), were associated with the stimulatory effect of dalteparin. Enoxaparin and dalteparin demonstrated opposing effects on inflammatory markers. These observed effects could be due to the presence of structurally different components in the two LMWHs arising from different methods of depolymerisation. This study provides a platform for further studies investigating the usefulness of enoxaparin in various inflammatory diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)