Merck
FOXM1 confers to epithelial-mesenchymal transition, stemness and chemoresistance in epithelial ovarian carcinoma cells.
Oncotarget (2014-12-30)
Wen-Tai Chiu, Yu-Fang Huang, Huei-Yu Tsai, Chien-Chin Chen, Chang-Hwa Chang, Soon-Cen Huang, Keng-Fu Hsu, Cheng-Yang Chou
ABSTRAKT
Chemoresistance to anti-cancer drugs substantially reduces survival in epithelial ovarian cancer. In this study, we showed that chemoresistance to cisplatin and paclitaxel induced the epithelial-mesenchymal transition (EMT) and a stem cell phenotype in ovarian cancer cells. Chemoresistance was associated with the downregulation of epithelial markers and the upregulation of mesenchymal markers, EMT-related transcription factors, and cancer stem cell markers, which enhanced invasion and sphere formation ability. Overexpression of FOXM1 increased cisplatin-resistance and sphere formation in cisplatin-sensitive and low FOXM1-expressing ovarian cancer cells. Conversely, depletion of FOXM1 via RNA interference reduced cisplatin resistance and sphere formation in cisplatin-resistant and high FOXM1-expressing cells. Overexpression of FOXM1 also increased the expression, nuclear accumulation, and activity of β-CATENIN in chemoresistant cells, whereas downregulation of FOXM1 suppressed these events. The combination of cisplatin and the FOXM1 inhibitor thiostrepton inhibited the expression of stem cell markers in chemoresistant cells and subcutaneous ovarian tumor growth in mouse xenografts. In an analysis of 106 ovarian cancer patients, high FOXM1 levels in tumors were associated with cancer progression and short progression-free intervals. Collectively, our findings highlight the importance of FOXM1 in chemoresistance and suggest that FOXM1 inhibitors may be useful for treatment of ovarian cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
USP
Transplatyna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human FOXM1
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Foxm1
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard