Merck
Self-assembled liquid-crystalline folate nanoparticles for in vitro controlled release of doxorubicin.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-02-11)
Rahul Misra, Sanat Mohanty
ABSTRAKT
Liquid-crystalline folate nanoparticles are ordered in structure which offers several advantages like high encapsulation of drugs, controlled release rates, biocompatible in nature. Moreover, it facilitates the cellular uptake of nanodrugs without any extra step of folate ligand based targeting. The size of these nanocarriers as well as the release profiles of drugs from these nano-carriers can be controlled precisely. Folate molecules self-assemble in ordered stacks and columns even at low concentration of 0.1wt%. Doxorubicin molecules get intercalated within the folate stacks and are developed into nanoparticles. These nanoparticles are composed of highly ordered folate self-assembly which encapsulate doxorubicin molecules. These drug molecules can be released in a controlled manner by disrupting this assembly in the environment of monovalent cations. The ordered structure of folate nanoparticles offers low drug losses of about 4-5%, which is significant in itself. This study reports the size-control method of forming doxorubicin encapsulated folate nanoparticles as well as the parameters to control the release rates of doxorubicin through liquid-crystalline folate nanoparticles. It has been demonstrated that doxorubicin release rates can be controlled by controlling the size of the nanoparticles, cross-linking cation and cross-linking concentration. The effect of different factors like drug loading, release medium, and pH of the medium on doxorubicin release rates was also studied. Moreover, this study also addresses the comparative in vitro cytotoxic performance of Doxorubicin loaded folate nanoparticles and cellular uptake of nano-carriers on cancer and normal cell line.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM