Merck
Genotoxicity testing of esterified propoxylated glycerol (EPG).
Regulatory toxicology and pharmacology : RTP (2014-12-17)
David H Bechtel
ABSTRAKT
Four versions of esterified propoxylated glycerols (EPGs) were evaluated for potential genotoxicity using a range of in vitro and in vivo assays. H-EPG-05 HR/SO 9:1, H-EPG-05 soyate, and H-EPG-14 soyate were non-mutagenic in reverse mutation assays (maximum concentration 1000 μg/plate) using Salmonella typhimurium and Escherichia coli. Heated and unheated H-EPG-05 HR/SO 9:1 and EPG-05 HR/ST 45:55 were likewise non-mutagenic in reverse mutation assays in S. typhimurium strains TA98 and TA100 (maximum concentration 5000 μg/plate). H-EPG-05 HR/SO 9:1, H-EPG-05 soyate, and H-EPG-14 soyate, were devoid of mutagenic activity in a mouse lymphoma assay in L5178Y tk +/- cells (maximum concentration 200 μg/plate for H-EPG-05 HR/SO 9:1; 100 μg/plate for H-EPG-05 soyate and H-EPG-14 soyate), and a chromosomal aberration test using human lymphocytes (maximum concentration 50 μg/plate for H-EPG-05 HR/SO 9:1 and H-EPG-05 soyate; 60 μg/plate for H-EPG-14 soyate). All assays were conducted with and without metabolic activation. Additionally, H-EPG-05 HR/SO 9:1, H-EPG-05 soyate, and H-EPG-14 soyate were non-genotoxic in unscheduled DNA synthesis tests in rats (maximum dose 2000 mg/kg). Based on the results of these assays it was concluded that these versions of EPG were not genotoxic under any of the conditions of the assays performed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glikol propylenowy, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
9-Aminoacridine, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Erucic acid, technical, ~90% (GC)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyrene, 98%
Supelco
Erucic acid, analytical standard
Supelco
Pyrene, analytical standard
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Erucic acid, ≥99% (capillary GC)
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
N-(Cyanomethyl)acetamide, ≥97.0% (elemental analysis)
Sigma-Aldrich
2-Nitrofluorene, 98%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.