Merck
Eyelid closure in embryogenesis is required for ocular adnexa development.
Investigative ophthalmology & visual science (2014-11-08)
Qinghang Meng, Maureen Mongan, Vinicius Carreira, Hisaka Kurita, Chia-Yang Liu, Winston W-Y Kao, Ying Xia
ABSTRAKT
Mammalian eye development requires temporary fusion of the upper and lower eyelids in embryogenesis. Failure of lid closure in mice leads to an eye open at birth (EOB) phenotype. Many genetic mutant strains develop this phenotype and studies of the mutants lead to a better understanding of the signaling mechanisms of morphogenesis. The present study investigates the roles of lid closure in eye development. Seven mutant mouse strains were generated by different gene ablation strategies that inactivated distinct signaling pathways. These mice, including systemic ablation of Map3k1 and Dkk2, ocular surface epithelium (OSE) knockout of c-Jun and Egfr, conditional knockout of Shp2 in stratified epithelium (SE), as well as the Map3k1/Jnk1 and Map3k1/Rhoa compound mutants, all exhibited defective eyelid closure. The embryonic and postnatal eyes in these mice were characterized by histology and immunohistochemistry. Some eye abnormalities, such as smaller lens in the Map3k1-null mice and Harderian gland hypoplasia in the Dkk2-null mice, appeared to be mutant strain-specific, whereas other abnormalities were seen in all mutants examined. The common defects included corneal erosion/ulceration, meibomian gland hypoplasia, truncation of the eyelid tarsal muscles, failure of levator palpebrae superioris (LPS) extension into the upper eyelid and misplacement of the inferior oblique (IO) muscle and inferior rectus (IR) muscle. The muscle defects were traced to the prenatal fetuses. In addition to providing a protective barrier for the ocular surface, eyelid closure in embryogenesis is required for the development of ocular adnexa, including eyelid and extraocular muscles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC