Merck
Hyperbaric pressure and increased susceptibility to glutamate toxicity in retinal ganglion cells in vitro.
Molecular vision (2014-05-16)
Makoto Aihara, Yi-Ning Chen, Saiko Uchida, Mao Nakayama, Makoto Araie
ABSTRAKT
To investigate the effect of hyperbaric pressure on purified retinal ganglion cells (RGCs) and the additive effect of hyperbaric pressure on glutamate-induced RGC death. An RGC primary culture from 8-day-old Wistar rats was prepared and cultured in a hyperbaric chamber. The RGC survival rate under various pressure conditions and with 5 or 25 µM of glutamate stimulation was determined and compared with that of RGCs under isobaric conditions. First, RGCs were cultured at atmospheric pressure (0 mmHg) and under hyperbaric pressure (+30 and +90 mmHg, with pressure fluctuations varying from 0 to +30 or +60 mmHg). Next, RGCs were cultured at +15, +30, and +90 mmHg with the addition of 5 or 25 µM of glutamate. The effects of N-Methyl-D-aspartic acid (NMDA) and 2-amino-3-(5-methyl-3-oxo-1,2- oxazol-4-yl)propanoic acid (AMPA)/kainate receptor antagonists, MK-801, and 6,7-dinitroquinoxaline-2,3-dione (DNQX), on cell survival were assessed. Additionally, types of cell death and the induction of Bcl-2-associated X protein (BAX) leading to apoptosis were studied under hyperbaric pressure conditions and/or with 5 µM of glutamate. RGC death was not induced under increasing or fluctuating pressure conditions. RGC death was induced by 25 µM of glutamate and increased as pressure increased. RGC death was not induced by 5 µM of glutamate but was induced by and increased with increasing pressure. MK-801 and DNQX significantly reduced glutamate-induced RGC death, and DNQX was more effective than MK-801. Under hyperbaric pressure conditions, the addition of 5 µM of glutamate resulted in the induction of apoptosis and BAX, which did not occur under hyperbaric pressure conditions or with the addition of glutamate alone. In a rat RGC culture, hyperbaric pressure alone did not induce RGC death but increased RGC susceptibility to glutamate toxicity, which may be of relevance to ocular diseases with pressure-induced RGC death.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
BDNF human, Carrier free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), suitable for cell culture
Supelco
Staurosporine, VETRANAL®, analytical standard
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Supelco
L-Lysine, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
DNQX, ≥98% (TLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Anti-Thy-1.1 Antibody, clone OX-7, clone OX-7, Chemicon®, from mouse
Supelco
Forskolin, analytical standard