Merck
Mechanism of back electron transfer in an intermolecular photoinduced electron transfer reaction: solvent as a charge mediator.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-07-22)
Sudhakar Narra, Yoshifumi Nishimura, Henryk A Witek, Shinsuke Shigeto
ABSTRAKT
Back electron transfer (BET) is one of the important processes that govern the decay of generated ion pairs in intermolecular photoinduced electron transfer reactions. Unfortunately, a detailed mechanism of BET reactions remains largely unknown in spite of their importance for the development of molecular photovoltaic structures. Here, we examine the BET reaction of pyrene (Py) and 1,4-dicyanobenzene (DCB) in acetonitrile (ACN) by using time-resolved near- and mid-IR spectroscopy. The Py dimer radical cation (Py2(·+)) and DCB radical anion (DCB(·-)) generated after photoexcitation of Py show asynchronous decay kinetics. To account for this observation, we propose a reaction mechanism that involves electron transfer from DCB(·-) to the solvent and charge recombination between the resulting ACN dimer anion and Py2(·+). The unique role of ACN as a charge mediator revealed herein could have implications for strategies that retard charge recombination in dye-sensitized solar cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pyrene, BCR®, certified reference material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,4-Dicyanobenzene, 98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%