Merck
  • High platelet reactivity is associated with vascular function in patients after percutaneous coronary intervention receiving clopidogrel.
High platelet reactivity is associated with vascular function in patients after percutaneous coronary intervention receiving clopidogrel.
International journal of cardiology (2014-12-17)
Gerasimos Siasos, Evangelos Oikonomou, Marina Zaromitidou, Stamatios Kioufis, Manolis Vavuranakis, Konstantinos Maniatis, Eleni Kokkou, Nikolaos Papageorgiou, Spyridon Papaioannou, Panagiotis Tourikis, Athanasios G Papavassiliou, Christodoulos Stefanadis, Dimitris Tousoulis
ABSTRAKT
In the present study, we evaluated the association of platelet reactivity with vascular function in patients after percutaneous coronary intervention receiving clopidogrel treatment. We enrolled 150 patients with stable CAD receiving clopidogrel regimen (75 mg/d), 1 month after percutaneous coronary intervention. Carotid-femoral pulse wave velocity (PWV) was measured as an index of aortic stiffness and augmentation index (AIx) as an index of arterial wave reflections. High on treatment platelet reactivity (HPR) was evaluated using VerifyNow Assay. VerifyNow reports its results in P2Y12 reaction units (PRU), and the diagnostic cutoff value is 230 PRU. Patients were evaluated prospectively up to 24 months. The primary end point was a composite of death from cardiovascular causes, nonfatal major cardiovascular events and hospitalization for cardiovascular causes. There was no difference in the basic clinical and demographic characteristics between subjects with HPR and non-HPR. Subjects with high on treatment platelet reactivity and PRU>230 had significantly increased PWV (8.81 ± 2.25 m/s vs. 7.69 ± 1.95 m/s, p = 0.001) and AIx (25.27 ± 8.67% vs. 20.87 ± 10.57%, p = 0.04) compared to subjects with PRU≤230. PWV was also associated with PRU (r = 0.23, p = 0.02). HPR was associated with significantly increased risk of primary end point [HR = 5.38, 95%CI:(1.15, 26.04), p = 0.03]. Increased platelet reactivity is associated with impaired arterial stiffness in patients after percutaneous coronary intervention receiving clopidogrel treatment, highlighting another clinical factor implicated in individual platelet response to antiplatelet therapy. Moreover, increased platelet reactivity is associated with adverse outcome in these patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
USP
Aspiryna, United States Pharmacopeia (USP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Aspirin, meets USP testing specifications
Informacje o cenach i dostępności nie są obecnie dostępne.