Merck
Direct recognition of the C-terminal polylysine residues of nonstop protein by Ltn1, an E3 ubiquitin ligase.
Biochemical and biophysical research communications (2014-10-12)
Kwang Hoon Sung, Hyun Kyu Song
ABSTRAKT
When mRNAs lack stop codons, errors in gene expression and coding of aberrant proteins that are harmful in cells can result. In Saccharomyces cerevisiae, a 180-kDa E3-ubiquitin ligase, Ltn1 has been known to associate with ribosomes and marks translationally-arrested aberrant nascent polypeptides for proteasomal degradation. Here, we demonstrate the Ltn1 E3-ubiquitin ligase directly binds to the nonstop proteins and efficiently ubiquitylates them. The middle domain of Ltn1 is responsible for recognizing the polylysine residues of the nonstop protein with an affinity of 2-3μM. This biochemical characterization of Ltn1 expands our knowledge regarding the fundamental process that removes aberrant nascent polypeptides in eukaryotes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Galaktoza, United States Pharmacopeia (USP) Reference Standard
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%