Merck
  • Optimization of a Raman microscopy technique to efficiently detect amorphous-amorphous phase separation in freeze-dried protein formulations.
Optimization of a Raman microscopy technique to efficiently detect amorphous-amorphous phase separation in freeze-dried protein formulations.
Journal of pharmaceutical sciences (2014-10-03)
Kelly M Forney-Stevens, Michael J Pelletier, Evgenyi Y Shalaev, Michael J Pikal, Robin H Bogner
ABSTRAKT
A confocal Raman microscopic technique was optimized to more efficiently detect amorphous-amorphous phase separation in freeze-dried protein formulations. A Renishaw Raman inVia confocal microscope was used to collect 100-200 μm line maps (2 μm step size) of freeze-dried protein-excipient formulations. At each point across the line map, the composition was evaluated from the intensity of the nonoverlapping peaks representative of each component. Collection aperture, scan time, and line map length significantly contributed to the phase-separation analysis, whereas different sample preparation methods did not affect the analysis. Using the optimized parameters (i.e., large aperture 5 s scan time, 200 μm line map), phase separation was successfully detected in binary polymer formulations and was comparable to the previously developed Raman method. However, the previous method required 2.5 h/sample, whereas the optimized method only requires 0.5 h/sample. Phase separation was detected in the following protein-excipient formulations: lysozyme-trehalose (1:1), lysozyme-isomaltose (1:1), β-lactoglobulin-dextran (1:1), β-lactoglobulin-dextran (1:3), and β-lactoglobulin-trehalose (1:1). Phase separation was not detected in lysozyme-sucrose (1:1) and β-lactoglobulin-sucrose (1:1) formulations. The optimized method successfully detected phase separation in several protein formulations, where phase separation was previously suspected, and promised to be a useful tool for detection of phase separation in amorphous therapeutic formulations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%