Merck
  • Detection of previously unknown fumonisin P analogue mycotoxins in a Fusarium verticillioides culture by high-performance liquid chromatography-electrospray ionization time-of-flight and ion trap mass spectrometry.
Detection of previously unknown fumonisin P analogue mycotoxins in a Fusarium verticillioides culture by high-performance liquid chromatography-electrospray ionization time-of-flight and ion trap mass spectrometry.
Journal of chromatographic science (2013-06-12)
Tibor Bartók, László Tölgyesi, Árpád Szécsi, Ákos Mesterházy, Mihály Bartók, Ernő Gyimes, Antal Véha
ABSTRAKT
Five previously unknown fumonisin mycotoxins (iso-FP1, iso-FP(2,3a), iso-FP(2,3b), FP4 and iso-FP4) and three previously described FP analogues (FP(1-3)) were detected in a solid rice culture inoculated with Fusarium verticillioides. The fumonisins were characterized by high-performance liquid chromatography-electrospray ionization time-of-flight mass spectrometry (HPLC-ESI-TOFMS) and ion trap mass spectrometry (ITMS). The FP isomers were separated by using a flat gradient on a special, high-coverage C18, narrow-bore HPLC column (YMC-Pack J'sphere ODS H80), which was suggested for the separation of structural isomers. The verified structures of the FP(1-3) mycotoxins, the relative retention times (by HPLC-ESI-TOFMS and ITMS), the exact masses of the molecular ions (by TOFMS) and the masses of the product ions, including the hydrocarbon backbones (by ITMS) of the new compounds, strongly suggested their structures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard