Merck
Platelet factor 4 limits Th17 differentiation and ischaemia-reperfusion injury after liver transplantation in mice.
Scandinavian journal of immunology (2014-12-03)
H Guo, Y Wang, Z Zhao, X Shao
ABSTRAKT
Liver ischaemia/reperfusion injury (IRI) is a serious pathologic process encountered in a number of clinical syndromes including liver transplantation, liver resection, trauma and haemorrhagic shock. Platelet factor 4 (PF4) was the first discovered CXC chemokine and is found in platelet granules at very high concentration. In this study, we provide strong evidence that PF4 is involved directly in liver innate immune response against IRI by regulating Th17 differentiation. PF4 deficiency aggravates liver IRI, as shown by higher serum alanine aminotransferase (ALT) levels and Suzuki scores. PF4 deficiency promotes Th17 response with higher levels of IL-23, IL-6 and IL-17, which aggravates liver IRI. Furthermore, PF4 deficiency limits suppressor of cytokine signalling 3 (SOCS3) expressions, and PF4 fails to suppress expression of IL-17 in cells transfected with SOCS3 SiRNA. In conclusion, PF4 limits liver IRI through IL-17 inhibition via upregulation of SOCS3.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
USP
Bromek cetrymonium, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent