Merck
Influence of lidocaine hydrochloride and penetration enhancers on the barrier function of human skin.
International journal of pharmaceutics (2014-10-12)
Kazumasa Hirata, Diar Mohammed, Jonathan Hadgraft, Majella E Lane
ABSTRAKT
Skin penetration enhancers (SPEs) are commonly employed in pharmaceutical and personal care products. These compounds transiently alter the barrier properties of the skin and we have previously investigated the effects of specific SPEs on skin barrier function in vivo. In the present study the effects of incorporation of an active pharmaceutical ingredient (API), lidocaine hydrochloride (LID HCl) in the SPEs previously studied were investigated. Solutions of LID HCl were prepared and applied to the volar forearm of human subjects with occlusion for 24h. Subsequently, tape stripping and trans epidermal water loss (TEWL) measurements were conducted for treated and control sites. The activities of the desquamatory proteases, kallikrein 5 (KLK 5) and kallikrein 7 (KLK 7) and API content were also measured from the tape strips. The propylene glycol (PG) formulation increased TEWL significantly (p<0.05) compared with the other SPEs and a mixture of the SPEs. However, only the isopropyl myristate (IPM) solution altered protease activity with a significant observed increase in kallikrein 5 (KLK 5). Incorporation of LID HCl appeared to ameliorate the effects of some of the SPEs on TEWL measurements compared with our previous study. Overall uptake of LID HCl into skin from the various formulations correlated very well with changes in TEWL. The findings should have implications for the choice of SPEs in topical and transdermal formulations, particularly where the skin barrier function of patients is already impaired for example in atopic eczema or psoriasis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
USP
Isopropyl myristate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%