Merck
  • Development and validation of a liquid-chromatography high-resolution tandem mass spectrometry approach for quantification of nine cytochrome P450 (CYP) model substrate metabolites in an in vitro CYP inhibition cocktail.
Development and validation of a liquid-chromatography high-resolution tandem mass spectrometry approach for quantification of nine cytochrome P450 (CYP) model substrate metabolites in an in vitro CYP inhibition cocktail.
Analytical and bioanalytical chemistry (2014-05-17)
Julia Dinger, Markus R Meyer, Hans H Maurer
ABSTRAKT
Knowledge about the cytochrome P450 (CYP) inhibition potential of new drug candidates is important for drug development because of its risk of interactions. For novel psychoactive substances (NPS), corresponding data are not available. For developing a general drug inhibition cocktail assay, a liquid-chromatography high-resolution tandem mass spectrometry multi-analyte approach was developed and validated for quantifying low concentrations of O-diethyl phenacetin for CYP 1A2, 7-hydroxy coumarin for CYP 2A6, 4-hydroxy bupropion for CYP 2B6, N-diethyl amodiaquine for CYP 2C8, 4-hydroxy diclofenac for CYP 2C9, 5-hydroxy omeprazole for CYP 2C19, O-dimethyl dextromethorphan for CYP 2D6, 6-hydroxy chlorzoxazone for CYP 2E1, and 6-beta-hydroxy testosterone for CYP 3A in the incubation mixture in the presence of substrates and inhibitors. The tested matrix effects ranged from 63 to 141 % and the recoveries from 95 to 110 %. Time-saving one-point calibration allowed sufficient quantification, although some of the validation results for 7-hydroxy coumarin, 4-hydroxy bupropion, 4-hydroxy diclofenac, and 6-beta-hydroxy testosterone were outside the acceptance criteria (AC) but without influence of the IC50 calculation. Validation showed also that the approach was sensitive and selective using mass spectral multiplexing. In conclusion, the presented assay was suitable for the quantification of the model substrate metabolites and could be used for the development of a CYP inhibition assay for testing most CYPs and a wide range of drugs of abuse.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Dextrorphan
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Supelco
Coumarin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Coumarin, primary reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Coumarin, European Pharmacopoeia (EP) Reference Standard