Merck
  • Transcriptome analysis by Illumina high-throughout paired-end sequencing reveals the complexity of differential gene expression during in vitro plantlet growth and flowering in Amaranthus tricolor L.
Transcriptome analysis by Illumina high-throughout paired-end sequencing reveals the complexity of differential gene expression during in vitro plantlet growth and flowering in Amaranthus tricolor L.
PloS one (2014-06-26)
Shengcai Liu, Huaqin Kuang, Zhongxiong Lai
ABSTRAKT
Amaranthus tricolor L. is a C4 plant, which is consumed as a major leafy vegetable in some tropical countries. Under conditions of high temperature and short daylight, Am. tricolor readily bolts and blooms, degrading leaf quality. A preliminary in vitro flowering study demonstrated that the flowering control pathway in Am. tricolor may differ from that of Arabidopsis. Nevertheless, no transcriptome analysis of the flowering process in Amaranthus has been conducted. To study Am. tricolor floral regulatory mechanisms, we conducted a large-scale transcriptome analysis--based on Illumina HiSeq sequencing of cDNA libraries generated from Am. tricolor at young seedling (YSS), adult seedling (ASS), flower bud (FBS), and flowering (FS) stages. A total of 99,312 unigenes were obtained. Using BLASTX, 43,088 unigenes (43.39%) were found to have significant similarity with accessions in Nr, Nt, and Swiss-Prot databases. Of these unigenes, 11,291 were mapped to 266 KEGG pathways. Further analysis of the four digital transcriptomes revealed that 735, 17,184, 274, and 206 unigenes were specifically expressed during YSS, ASS, FBS, and FS, respectively, with 59,517 unigenes expressed throughout the four stages. These unigenes were involved in many metabolic pathways related to in vitro flowering. Among these pathways, 259 unigenes were associated with ubiquitin-mediated proteolysis, indicating its importance for in vitro flowering in Am. tricolor. Other pathways, such as circadian rhythm and cell cycle, also had important roles. Finally, 26 unigenes were validated by qRT-PCR in samples from Am. tricolor at YSS, ASS, FBS, and FS; their differential expressions at the various stages indicate their possible roles in Am. tricolor growth and development, but the results were somewhat similar to Arabidopsis. Because unigenes involved in many metabolic pathways or of unknown function were revealed to regulate in vitro plantlet growth and flowering in Am. tricolor, the process appears to be highly complex in this species.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O