Merck
Paraoxonase 1 and dietary hyperhomocysteinemia modulate the expression of mouse proteins involved in liver homeostasis.
Acta biochimica Polonica (2014-10-28)
Joanna Suszyńska-Zajczyk, Hieronim Jakubowski
ABSTRAKT
Homocysteine (Hcy), a product of methionine metabolism, is elevated by the consumption of a high-methionine diet that can cause fatty liver disease. Paraoxonase 1 (Pon1), a hydrolase expressed mainly in the liver and carried in the circulation on high-density lipoprotein, participates in Hcy metabolism. Low Pon1 activity is linked to fatty liver disease. We hypothesize that hyperhomocysteinemia and low Pon1 induce changes in gene expression that could impair liver homeostasis. To test this hypothesis, we analyzed the liver proteome of Pon1(-/-) and Pon1(+/+) mice fed a high methionine diet (1% methionine in the drinking water) for 8 weeks using 2D IEF/SDS-PAGE gel electrophoresis and MALDI-TOF mass spectrometry. We identified seven liver proteins whose expression was significantly altered in Pon1(-/-) mice. In animals fed with a control diet, the expression of three liver proteins involved in lipoprotein metabolism (ApoE), iron metabolism (Ftl), and regulation of nitric oxide generation (Ddah1) was up-regulated by the Pon1(-/-) genotype. In mice fed with a high-methionine diet, expression of four liver proteins was up-regulated and of three proteins was down-regulated by the Pon1(-/-) genotype. The up-regulated proteins are involved in lipoprotein metabolism (ApoE), energy metabolism (Atp5h), oxidative stress response (Prdx2), and nitric oxide regulation (Ddah1). The down-regulated proteins are involved in energy metabolism (Gamt), iron metabolism (Ftl), and catechol metabolism (Comt). Expression of one protein (Ftl) was up-regulated both by the Pon1(-/-) genotype and a high-methionine diet. Our findings suggest that Pon1 interacts with diverse cellular processes - from lipoprotein metabolism, nitric oxide regulation, and energy metabolism to iron transport and antioxidant defenses - that are essential for normal liver homeostasis and modulation of these interactions by a high-methionine diet may contribute to fatty liver disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)