Merck
Antimicrobial and anti-biofilm activity of thiourea derivatives incorporating a 2-aminothiazole scaffold.
Chemical & pharmaceutical bulletin (2015-03-12)
Joanna Stefanska, Grażyna Nowicka, Marta Struga, Daniel Szulczyk, Anna Eugenia Koziol, Ewa Augustynowicz-Kopec, Agnieszka Napiorkowska, Anna Bielenica, Wojciech Filipowski, Anna Filipowska, Aleksandra Drzewiecka, Gabriele Giliberti, Silvia Madeddu, Stefano Boi, Paolo La Colla, Giuseppina Sanna
ABSTRAKT
A series of new thiourea derivatives of 1,3-thiazole have been synthesized. All obtained compounds were tested in vitro against a number of microorganisms, including Gram-positive cocci, Gram-negative rods and Candida albicans. Compounds were also tested for their in vitro tuberculostatic activity against the Mycobacterium tuberculosis H37Rv strain, as well as two 'wild' strains isolated from tuberculosis patients. Compounds 3 and 9 showed significant inhibition against Gram-positive cocci (standard strains and hospital strain). The range of MIC values is 2-32 µg/mL. Products 3 and 9 effectively inhibited the biofilm formation of both methicillin-resistant and standard strains of S. epidermidis. The halogen atom, especially at the 3rd position of the phenyl group, is significantly important for this antimicrobial activity. Moreover, all obtained compounds resulted in cytotoxicity and antiviral activity on a large set of DNA and RNA viruses, including Human Immunodeficiency Virus type 1 (HIV-1) and other several important human pathogens. Compound 4 showed activity against HIV-1 and Coxsackievirus type B5. Seven compounds resulted in cytotoxicity against MT-4 cells (CC50<10 µM).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard