Merck
  • Identification of indicator components for the discrimination of Cassia plants in health teas and development of analytical method for the components.
Identification of indicator components for the discrimination of Cassia plants in health teas and development of analytical method for the components.
Journal of AOAC International (2014-08-26)
Mitsuko Takahashi, Katsumi Sakurai, Hisashi Fujii, Koichi Saito
ABSTRAKT
Components that could be used as indicators for the discrimination of senna (Cassia angustifolia) from other cassia plants contained in health teas were identified, and an analytical method for the components was developed. Our results revealed two components in senna that were not found in other Cassia spp. widely used in health teas, such as C. alata, C. corymbosa, C. obtusifolia, and C. occidentalis. Structural elucidation of the two components showed that they were isorhamnetin-3-O-gentiobioside and tinnevellin glucoside. We analyzed commercial health teas using the HPLC method developed in this study. The two indicator components were detected at 366 nm using an RP C18 column and gradient elution with a mixture of water and acetonitrile (with formic acid), as the mobile phase. Our analytical method by HPLC enabled the differentiation of senna from other Cassia plants present in health teas in which sennosides A and B were detected. Moreover, this method allowed us to predict the parts of senna in health teas from the amounts of isorhamnetin-3-O-gentiobioside and tinnevellin glucoside contained in the teas.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Wodorowęglan sodu, −40-+140 mesh, ≥95%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Methanol, analytical standard