Merck
Greater inflammatory activity and blunted glucocorticoid signaling in monocytes of chronically stressed caregivers.
Brain, behavior, and immunity (2014-09-23)
Gregory E Miller, Michael L M Murphy, Rosemary Cashman, Roy Ma, Jeffrey Ma, Jesusa M G Arevalo, Michael S Kobor, Steve W Cole
ABSTRAKT
Chronic stress is associated with morbidity and mortality from numerous conditions, many of whose pathogenesis involves persistent inflammation. Here, we examine how chronic stress influences signaling pathways that regulate inflammation in monocytes. The sample consisted of 33 adults caring for a family member with glioblastoma and 47 controls whose lives were free of major stressors. The subjects were assessed four times over eight months. Relative to controls, caregivers' monocytes showed increased expression of genes bearing response elements for nuclear-factor kappa B, a key pro-inflammatory transcription factor. Simultaneously, caregivers showed reduced expression of genes with response elements for the glucocorticoid receptor, a transcription factor that conveys cortisol's anti-inflammatory signals to monocytes. Transcript origin analyses revealed that CD14+/CD16- cells, a population of immature monocytes, were the predominate source of inflammatory gene expression among caregivers. We considered hormonal, molecular, and functional explanations for caregivers' decreased glucocorticoid-mediated transcription. Across twelve days, the groups displayed similar diurnal cortisol profiles, suggesting that differential adrenocortical activity was not involved. Moreover, the groups' monocytes expressed similar amounts of glucocorticoid receptor protein, suggesting that differential receptor availability was not involved. In ex vivo studies, subjects' monocytes were stimulated with lipopolysaccharide, and caregivers showed greater production of the inflammatory cytokine interleukin-6 relative to controls. However, no group differences in functional glucocorticoid sensitivity were apparent; hydrocortisone was equally effective at inhibiting cytokine production in caregivers and controls. These findings may help shed light on the mechanisms through which caregiving increases vulnerability to inflammation-related diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard