Merck
Enteric polymer based on pH-responsive aliphatic polycarbonate functionalized with vitamin E to facilitate oral delivery of tacrolimus.
Biomacromolecules (2015-02-26)
Menglin Wang, Jin Sun, Yinglei Zhai, He Lian, Cong Luo, Lin Li, Yuqian Du, Dong Zhang, Wenya Ding, Shuhong Qiu, Yuhai Liu, Longfa Kou, Xiangfei Han, Rongwu Xiang, Yongjun Wang, Zhonggui He
ABSTRAKT
To improve the bioavailability of orally administered drugs, we synthesized a pH-sensitive polymer (poly(ethylene glycol)-poly(2-methyl-2-carboxyl-propylene carbonate)-vitamin E, mPEG-PCC-VE) attempting to integrate the advantages of enteric coating and P-glycoprotein (P-gp) inhibition. The aliphatic polycarbonate chain was functionalized with carboxyl groups and vitamin E via postpolymerization modification. Optimized by comparison and central composite design, mPEG113-PCC32-VE4 exhibited low critical micelle concentration of 1.7 × 10(-6) mg/mL and high drug loading ability for tacrolimus (21.2% ± 2.7%, w/w). The pH-responsive profile was demonstrated by pH-dependent swelling and in vitro drug release. Less than 4.0% tacrolimus was released under simulated gastric fluid after 2.5 h, whereas an immediate release was observed under simulated intestinal fluid. The mPEG113-PCC32-VE4 micelles significantly increased the absorption of P-gp substrate tacrolimus in the whole intestine. The oral bioavailability of tacrolimus micelles was 6-fold higher than that of tacrolimus solution in rats. This enteric polymer therefore has the potential to become a useful nanoscale carrier for oral delivery of drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining