Merck
  • Porous hydrogel of wool keratin prepared by a novel method: an extraction with guanidine/2-mercaptoethanol solution followed by a dialysis.
Porous hydrogel of wool keratin prepared by a novel method: an extraction with guanidine/2-mercaptoethanol solution followed by a dialysis.
Materials science & engineering. C, Materials for biological applications (2014-07-27)
Yuki Ozaki, Yusuke Takagi, Hideki Mori, Masayuki Hara
ABSTRAKT
In this study, we show a novel simple method to prepare a sponge-like porous keratin hydrogel through the extraction of wool keratin in a solution containing guanidine hydrochloride and 2-mercaptoethanol followed by dialysis for both aggregation of keratin and recrosslink. The gel had a highly porous structure and a fast-swelling property in rehydration after freeze-drying. It had also high mechanical strength both in the tensile test and the measurement of dynamic viscoelasticity. Three types of animal cells, PC12 cells, HOS cells and murine embryonic fibroblasts, well attached and grew on the surface of the porous hydrogel.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%