Merck
Dead bacteria reverse antibiotic-induced host defense impairment in burns.
Journal of the American College of Surgeons (2014-09-23)
Lee-Wei Chen, Pei-Hsuan Chen, Chang-Phone Fung, Ching-Mei Hsu
ABSTRAKT
Burn patients can incur high rates of hospital-acquired infections. The mechanism of antibiotic exposure on inducing infection vulnerability has not been determined. This study aimed to examine the effects of antibiotic treatment on host defense mechanisms. First we treated C57/BL6 mice with combined antibiotic treatment after 30% to 35% total body surface area burn. Animals were sacrificed at 48 hours after sham or thermal injury treatment. Bacterial counts in intestinal lumen and mucosa were measured. Next, we treated animals with or without oral dead Escherichia coli or Staphylococcus aureus supplementation to stimulate Toll-like receptor in the intestinal mucosa. Toll-like receptor 4, antibacterial protein expression, nuclear factor (NF)-κB DNA-binding activity, and bacteria-killing activity in the intestinal mucosa; intestinal permeability; bacterial translocation to mesenteric lymph nodes; Klebsiella pneumoniae translocation; interleukin-6 in the blood; and phagocytic activity of alveolar macrophages, were assessed. Thermal injury increased microflora and NF-κB DNA-binding activity of the intestine. Systemic antibiotic treatment decreased gut microflora and increased bacterial translocation to mesenteric lymph nodes, intestinal permeability, and interleukin-6 levels in the blood. Antibiotic treatment also decreased bacteria-killing activity in intestinal mucosa and phagocytic activity of alveolar macrophages. Oral dead E coli and S aureus supplementation induced NF-κB DNA-binding activity, Toll-like receptor 4, and antibacterial protein expression of the intestinal mucosa. Taken together with the fact that dead bacteria reversed antibiotic-induced K pneumoniae translocation and intestinal and pulmonary defense impairment, we conclude that combined antibiotic treatment results in systemic host defense impairment in burns through the decrease in intestinal flora. We suggest that dead bacteria supplementation could induce nondefensin protein expression and reverse antibiotic-induced gut and lung defense impairment in burn patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Metronidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, free acid