Merck
  • IL-6 upregulates a disintegrin and metalloproteinase with thrombospondin motifs 2 (ADAMTS-2) in human osteosarcoma cells mediated by JNK pathway.
IL-6 upregulates a disintegrin and metalloproteinase with thrombospondin motifs 2 (ADAMTS-2) in human osteosarcoma cells mediated by JNK pathway.
Molecular and cellular biochemistry (2014-04-23)
Meltem Alper, Feray Kockar
ABSTRAKT
ADAMTS-2 and ADAMTS-3 (a disintegrin and metalloproteinase with thrombospondin type 1 motif 2) belong to the procollagen aminoproteinase subfamily of ADAMTS proteases. They play crucial roles in the collagen metabolism. To understand the regulation of ADAMTS-2 gene expression in osteoblastic cells, we have cloned a functional 760 bp of human ADAMTS-2 promoter. Sequence analysis of the ADAMTS-2 promoter region showed the absence of a TATA box, but identified a GC box, a CpG island, several GAGA boxes and several transcriptional factor binding sites, which may be valuable in the regulation of ADAMTS-2 transcription. We also elucidated that Interleukin 6 (IL-6) increases ADAMTS-2 and ADAMTS-3 mRNA and protein levels in different osteosarcoma cell lines namely, MG-63 and Saos-2. IL-6 also increases the transcriptional activation of the ADAMTS-2 gene promoter. Pathway inhibition studies revealed that ADAMTS-2 upregulation by IL-6 was mediated by JNK pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material