Merck
Structure-retention behaviour of biologically active fused 1,2,4-triazinones--correlation with in silico molecular properties.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-12-22)
Małgorzata Sztanke, Tomasz Tuzimski, Małgorzata Janicka, Krzysztof Sztanke
ABSTRAKT
The chromatographic behaviour and significant lipophilicity/hydrophobicity indices (log k(w), S, φ(0)) are presented for 21 biologically active fused 1,2,4-triazinones based on the linear relationship: log k = log k(w)-Sφ established for the retention on LC-18 HPLC column, using as mobile phases mixtures of three organic modifiers with water. The effect of these mobile phase modifiers on the chromatographic behaviour of solutes was established and the organic modifier of choice is suggested. The complex correlation of slopes versus intercepts obtained for acetonitrile, contrary to linear ones obtained for methanol and dioxane are disclosed. The observed difference in retention mechanism for acetonitrile compared to methanol and dioxane is explained by intermolecular interactions encoded in lipophilicity. Linear correlations with statistically significant levels between log kw values determined from three different chromatographic systems were obtained. The relationships between log k(w) constants (derived from the linear model for methanol-water mobile phases) and predicted log P and log S values by the use of various computational methods were investigated and these were established with high correlation coefficients. The predicted log P values plotted against φ(0 (MeOH)) indices showed the best fit. Principal component analysis was used to compare various lipophilicity parameters of the solutes and their in silico biological descriptors relevant to optimal pharmacokinetics profile. The similarities and dissimilarities between all the variables and molecular structures of solutes are presented. Statistically significant correlations were found between the chromatographic lipophilicity indices and the calculated pharmacokinetic descriptors: fraction unbound in brain (f(u, brain)), oral bioavailability (%F), permeability and intestinal absorption in jejunum (Caco-2), skin permeation (log K(p)) and blood/brain concentration (log BB).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Supelco
1,4-Dioxane, analytical standard
Supelco
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%