Merck
Establishment of optimized in vitro assay methods for evaluating osteocyte functions.
Journal of bone and mineral metabolism (2014-01-02)
Masashi Honma, Yuki Ikebuchi, Yoshiaki Kariya, Hiroshi Suzuki
ABSTRAKT
Recent studies have revealed that osteocytes play multiple important physiological roles. To analyze osteocyte functions in detail, an in vitro experimental system for primary osteocytes would be useful. Unfortunately, osteocytes tend to dedifferentiate and acquire osteoblast-like features even when the cells are cultured in three-dimensional (3D) collagen gel. Therefore, it is desirable to establish osteocyte culture conditions that prevent dedifferentiation over longer periods. In this study, we obtained systematic information about the influence of culture conditions on osteocyte differentiation states. Fetal bovine serum (FBS) concentrations from 0.1 to 0.5 % in 3D culture matrix did not significantly influence the expression of osteocyte markers. On the other hand, addition of Matrigel to the culture matrix significantly enhanced the expression of Rankl and late osteocyte markers such as Sost and Fgf23. Matrigel addition also inhibited upregulation of Opg and early osteocyte markers such as Dmp1 and Gp38. These effects on osteocyte properties were maximal at a Matrigel culture matrix content of 50 %. Matrigel addition to the matrix also increased dendritic process extension by osteocytes. In addition, Matrigel addition significantly stimulated tartrate-resistant acid phosphatase activity in co-culture with bone marrow macrophages. Among the conditions tested, 50 % Matrigel and 0.2 % FBS in type I collagen matrix were optimal for culture of primary osteocytes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Tartaric Acid, Pharmaceutical Secondary Standard; Certified Reference Material