Merck
Persistently altered epigenetic marks in the mouse uterus after neonatal estrogen exposure.
Molecular endocrinology (Baltimore, Md.) (2013-09-05)
Wendy N Jefferson, Dominique M Chevalier, Jazma Y Phelps, Amy M Cantor, Elizabeth Padilla-Banks, Retha R Newbold, Trevor K Archer, H Karimi Kinyamu, Carmen J Williams
ABSTRAKT
Neonatal exposure to diethylstilbestrol (DES) causes permanent alterations in female reproductive tract gene expression, infertility, and uterine cancer in mice. To determine whether epigenetic mechanisms could explain these phenotypes, we first tested whether DES altered uterine expression of chromatin-modifying proteins. DES treatment significantly reduced expression of methylcytosine dioxygenase TET oncogene family, member 1 (TET1) on postnatal day 5; this decrease was correlated with a subtle decrease in DNA 5-hydroxymethylcytosine in adults. There were also significant reductions in histone methyltransferase enhancer of zeste homolog 2 (EZH2), histone lysine acetyltransferase 2A (KAT2A), and histone deacetylases HDAC1, HDAC2, and HDAC3. Uterine chromatin immunoprecipitation was used to analyze the locus-specific association of modified histones with 2 genes, lactoferrin (Ltf) and sine oculis homeobox 1 (Six1), which are permanently upregulated in adults after neonatal DES treatment. Three histone modifications associated with active transcription, histone H3 lysine 9 acetylation (H3K9ac), H3 lysine 4 trimethylation (H3K4me3), and H4 lysine 5 acetylation (H4K5ac) were enriched at specific Ltf promoter regions after DES treatment, but this enrichment was not maintained in adults. H3K9ac, H4K5ac, and H3K4me3 were enriched at Six1 exon 1 immediately after neonatal DES treatment. As adults, DES-treated mice had greater differences in H4K5ac and H3K4me3 occupancy at Six1 exon 1 and new differences in these histone marks at an upstream region. These findings indicate that neonatal DES exposure temporarily alters expression of multiple chromatin-modifying proteins and persistently alters epigenetic marks in the adult uterus at the Six1 locus, suggesting a mechanism for developmental exposures leading to altered reproductive function and increased cancer risk.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-SIX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Przeciwciało anty-histonowe H3, CT, pan, klon A3S, królicze monoklonalne, culture supernatant, clone A3S, Upstate®
Sigma-Aldrich
Przeciwciało anty-acetylo-histonowe H4, serum, Upstate®
Sigma-Aldrich
Przeciwciało przeciwko acetylo-histonie H3 (Lys9), serum, Upstate®
Sigma-Aldrich
Przeciwciało przeciw dimetylo-histonowi H3 (Lys9), Upstate®, from rabbit