Merck
Effect of Monascus aged vinegar on isoflavone conversion in soy germ by soaking treatment.
Food chemistry (2015-05-16)
Ji-Cheng Chen, Jie Wang, Zhi-Jiang Wang, Yan-Jie Li, Jie Pang, He-Tong Lin, Shou-Wei Yin
ABSTRAKT
Soy germ rich in isoflavones has attracted much attention for health-promoting characteristics. An effective approach via Monascus aged vinegar soaking was adopted to enhance the aglycone amount. The profiles and interconversion of soy germ isoflavones via Monascus aged vinegar soaking were investigated, and the distribution in vinegars were also explored. The aglycones were dramatically increased by 40.76 times. Concomitantly, β-glycosides and malonylglycosides were significantly decreased. The proportion of aglycones presented a sharp increase with the endogenous β-glucosidase activity at the initial 4h incubation. There appeared to be correlations between β-glucosidase activity and the hydrolysis of conjugated isoflavones. The results demonstrated that the reactions of decarboxylation, de-esterification and de-glycosylation were involved in the Monascus aged vinegar soaking, supporting synergistic effects of enzymolysis by endogenous β-glucosidase from soy germ and acid hydrolysis of vinegars. Soaking by vinegar is a promising pathway for preparing aglycone-rich soy germ.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Genistin, from Glycine max (soybean), ≥95% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder