Merck
Optimal fusion of antibody binding domains resulted in higher affinity and wider specificity.
Journal of bioscience and bioengineering (2015-04-26)
Jinhua Dong, Tomoki Kojima, Hiroyuki Ohashi, Hiroshi Ueda
ABSTRAKT
Antibody is a very important protein in biotechnological and biomedical fields because of its high affinity and specificity to various antigens. Due to the rise of human antibody therapeutics, its cost-effective purification is an urgent issue for bio-industry. In this study, we made novel fusion proteins PAxPG with a flexible (DDAKK)n linker between the two Ig binding domains derived from Staphylococcus protein A and Streptococcus protein G. The fusion proteins bound human and mouse IgGs and their fragments with up to 58-times higher affinity and wider specificity than the parental binding domains. Interestingly, the optimal linker for human Fab fragment was n = 4, which was close to the modeled distance between the termini of domains bound to heavy chain, implying increased avidity as a possible mechanism. For binding to Fc, the longest n=6 linker gave the highest affinity, implying longer interchain distance between the two binding sites. The novel fusion protein with optimized interdomain linker length will be a useful tool for the purification and detection of various IgGs including mouse IgG1 that binds only weakly to natural protein A.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Maleimide, 99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
1,3-Propanediol, 98%
Informacje o cenach i dostępności nie są obecnie dostępne.