Merck
  • D-2-Hydroxyglutarate does not mimic all the IDH mutation effects, in particular the reduced etoposide-triggered apoptosis mediated by an alteration in mitochondrial NADH.
D-2-Hydroxyglutarate does not mimic all the IDH mutation effects, in particular the reduced etoposide-triggered apoptosis mediated by an alteration in mitochondrial NADH.
Cell death & disease (2015-03-27)
K Oizel, C Gratas, A Nadaradjane, L Oliver, F M Vallette, C Pecqueur
ABSTRAKT
Somatic mutations in isocitrate dehydrogenase (IDH)-1 and -2 have recently been described in glioma. This mutation leads to a neomorphic enzymatic activity as the conversion of isocitrate to alpha ketoglutarate (αKG) is replaced by the conversion of αKG to D-2-hydroxyglutarate (D-2HG) with NADPH oxidation. It has been suggested that this oncometabolite D-2HG via inhibition of αKG-dioxygenases is involved in multiple functions such as epigenetic modifications or hypoxia responses. The present study is aimed at deciphering how the mutant IDH can affect cancer pathogenesis, in particular with respect to its associated oncometabolite D-2HG. We show that the overexpression of mutant IDH in glioma cells or treatment with D-2HG triggered an increase in cell proliferation. However, although mutant IDH reduced cell sensitivity to the apoptotic inducer etoposide, D-2HG exhibited no effect on apoptosis. Instead, we found that the apoptotic effect was mediated through the mitochondrial NADH pool reduction and could be inhibited by oxamate. These data show that besides D-2HG production, mutant IDH affects other crucial metabolite pools. These observations lead to a better understanding of the biology of IDH mutations in gliomas and their response to therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis