Merck
  • Herb-drug interactions between Scutellariae Radix and mefenamic acid: Simultaneous investigation of pharmacokinetics, anti-inflammatory effect and gastric damage in rats.
Herb-drug interactions between Scutellariae Radix and mefenamic acid: Simultaneous investigation of pharmacokinetics, anti-inflammatory effect and gastric damage in rats.
Journal of ethnopharmacology (2015-05-20)
Sophia Yui Kau Fong, Yin Cheong Wong, Chen Xie, Zhong Zuo
ABSTRAKT
Scutellariae Radix (SR), the dried root of Scutellariae baicalensis Georgi, has a lot in common with non-steroidal anti-inflammatory drugs (NSAIDs). Their similarities in therapeutic action (anti-inflammation) and metabolic pathways (phase II metabolisms) may lead to co-administration by patients with the potential of pharmacokinetic and/or pharmacodynamic interactions. The current study aims to investigate the potential interactions between SR and an NSAID, mefenamic acid (MEF), on the overall pharmacokinetic dispositions, anti-inflammatory effects and adverse effects in rats. The current study simultaneously monitored the pharmacokinetic and pharmacodynamic interactions in a single animal. Four groups of Sprague-Dawley rats (n=7 each) received oral doses of a standardized SR extract (300mg/kg, twice daily), MEF (40mg/kg, daily), combination of SR extract and MEF, and vehicle control, respectively, for 5 days. On Day 5, blood samples were collected after first dose over 24h for the determination of (1) plasma concentrations of SR bioactive components, MEF and its metabolites by LC-MS/MS, and (2) prostaglandin E2 (PGE2) production and cyclooxygenase-2 (COX-2) gene expression by ex vivo analyses using LPS-stimulated RAW264.7 macrophage cells, ELISA and real time-PCR. After the rats were sacrificed, stomachs were isolated to assess their gross mucosal damage. Statistical comparisons were conducted using ANOVA and t-test. Minimal pharmacokinetic interaction between SR extract and MEF was observed. Co-administration of SR extract and MEF did not significantly alter the plasma concentration-time profile or the pharmacokinetic parameters such as Cmax, AUC0→24, Tmax or clearance. Pharmacodynamic interaction via the COX-2 pathway was observed. The PGE2 level in LPS-stimulated RAW264.7 cells treated with plasma collected from control group over the 24h sampling (AUC0→24[PGE2]) was 191981±8789pg/mlhr, which was significantly reduced to 174,780±6531 and 46,225±1915pg/mlhr by plasma collected from rats administered with SR extract and MEF, respectively. Co-administration of SR extract and MEF further potentiated the PGE2 inhibition, with an AUC0→24[PGE2] of 37013±2354pg/mlhr (p<0.05, compared to SR or MEF group). By analyzing the COX-2 gene expression, SR extract significantly prolonged the COX-2 inhibitory effect of MEF over the 24h (p<0.05). Furthermore, the MEF-induced stomach ulcer after the 5-day treatment, as evidenced by the increased gross ulcer index and sum of lesion length (p<0.05, compared to control), could be alleviated by co-administration with SR extract (p<0.05). Co-administration of SR extract and MEF potentiated the anti-inflammatory effects, alleviated the MEF-induced stomach adverse effect while having minimal pharmacokinetic interactions. Our findings provide insight for combination therapy of SR extract and MEF against inflammatory diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)