Merck
Expanded corn starch as a versatile material in atom transfer radical polymerization (ATRP) of styrene and methyl methacrylate.
Carbohydrate polymers (2015-06-17)
Ankushi Bansal, Arvind Kumar, Patnam Padma Latha, Siddharth Sankar Ray, Alok Kumar Chatterjee
ABSTRAKT
Polymerization of styrene (St) and methyl methacrylate (MMA) was performed by surface initiated (SI) and activator generated by electron transfer (AGET) systems of atom transfer radical polymerization (ATRP) using renewable expanded corn starch (ECS) as a support. This prepared ECS is found to have V type crystallinity with 50 m(2)g(-1) surface area (<1m(2)g(-1) for corn starch (CS)) and average pore volume of 0.43 cm(3)g(-1) (<0.1cm(3)g(-1) for CS). In SI-ATRP, hydroxyl groups on ECS were converted into macro-initiator by replacing with 2-bromoisobutyryl bromide (BIBB) with a 0.06 degree of substitution determined from NMR. In AGET-ATRP, CuBr2/ligand complex get adsorbed on ECS (Cu(II)/ECS=10 wt.%) to catalyze the polymerization. Synthesized PS/PMMA was characterized by SEM, FT-IR, (1)H NMR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%