Merck
Efficient charge separation based on type-II g-C3N4/TiO2-B nanowire/tube heterostructure photocatalysts.
Dalton transactions (Cambridge, England : 2003) (2015-06-24)
Hongmei Chen, Yinghao Xie, Xiaoqin Sun, Meilin Lv, Fangfang Wu, Lei Zhang, Li Li, Xiaoxiang Xu
ABSTRAKT
Separation of photo-generated charges has played a crucial role in controlling the actual performance of a photocatalytic system. Here we have successfully fabricated g-C3N4/TiO2-B nanowire/tube heterostructures through facile urea degradation reactions. Owing to the effective separation of photo-generated charges associated with the type-II band alignment and intimate interfacial contacts between g-C3N4 and TiO2-B nanowires/tubes, such heterostructures demonstrate an improved photocatalytic activity over individual moieties. Synthetic conditions such as hydrothermal temperatures for the preparation of TiO2-B and the weight ratio of TiO2-B to urea were systematically investigated. A high crystallinity of TiO2-B as well as the proper growth of g-C3N4 on its surface are critical factors for a better performance. Our simple synthetic method and the prolonged lifetime of photo-generated charges signify the importance of type-II heterostructures in the photocatalytic applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O